Product Name
3-Methyl-3-buten-1-ol 97%
Product Number
129402
Product Brand
aldrich
CAS Number
763-32-6
Molecular Weight
86.13
Test
Specification
APPEARANCE (COLOR)Colorless to Faint Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE