Product Name
2,4,6-Trimethylaniline 98%
Product Number
132179
Product Brand
aldrich
CAS Number
88-05-1
Molecular Weight
135.21
Test
Specification
APPEARANCE (COLOR)Colorless to Very Dark Yellow and Colorless to Very Dark Brown-Yellow and Colorless to Very Dark Brown
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE