Product Name
2-Methyl-2-oxazoline 98%
Product Number
137448
Product Brand
aldrich
CAS Number
1120-64-5
Molecular Weight
85.10
Test
Specification
APPEARANCE (COLOR)Colorless to Faint Yellow
APPEARANCE (FORM)Liquid
TITRATION (NT) HCLO4 0.1M97.5 - 102.5 %
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE