Product Name
2-Ethyl-2-oxazoline ≥99%
Product Number
137456
Product Brand
aldrich
CAS Number
10431-98-8
Molecular Weight
99.13
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE