Product Name
Glycidyl 4-methoxyphenyl ether 99%
Product Number
148067
Product Brand
aldrich
CAS Number
2211-94-1
Molecular Weight
180.20
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Powder with Chunk(s)
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE