Product Name
Boc-Aib-OH ≥99.0% (T)
Product Number
15466
Product Brand
aldrich
CAS Number
30992-29-1
Molecular Weight
203.24
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) NAOH 0.1M99.0 - 101.0 %
PURITY (TLC AREA %)≥ 99.0 %
MELTING POINT118 - 122 C
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)0.5G IN 10ML METHANOL
CARBON CONTENT53.19 % (THEORY)
HYDROGEN CONTENT8.43 % (THEORY)
NITROGEN CONTENT6.89 % (THEORY)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE