Product Name
(Chloromethyl)cyclopropane 97%
Product Number
184667
Product Brand
aldrich
CAS Number
5911-08-0
Molecular Weight
90.55
Test
Specification
Appearance (Color)Colorless
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (GC)≥96.5 %