Product Name
trans-7-Tetradecene 97%
Product Number
227560
Product Brand
aldrich
CAS Number
41446-63-3
Molecular Weight
196.37
Test
Specification
APPEARANCECOLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE AND STANDARD AS
 ILLUSTRATED ON PAGE 20C OF EDITION I,
 VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR
 SPECTRA".
GAS LIQUID96.5% (MINIMUM)
 REVISED OCTOBER 18, 1999 JSB