Product Name
(R,R)-(+)-Hydrobenzoin 99%, optical purity ee: 99% (HPLC)
Product Number
256277
Product Brand
aldrich
CAS Number
52340-78-0
Molecular Weight
214.26
Test
Specification
Appearance (Color)White to Beige
Appearance (Form)Crystal to Powder
Infrared spectrumConforms to Structure
Optical Rotation89 - 95 deg
Enantiomeric Excess≥98.5 %
GC (area %)≥98.5 %