Product Name
Iridium wire, diam. 0.15 mm, 99.9% trace metals basis
Product Number
266841
Product Brand
aldrich
CAS Number
7439-88-5
Molecular Weight
192.22
Test
Specification
APPEARANCESHINY SILVER OR GREY WIRE
 0.15 MM IN DIAMETER
ICP ASSAYCONFIRMS IRIDIUM COMPONENT.
TRACE ANALYSIS,2000 PPM (MAXIMUM) TOTAL METALLIC IMPURITIES
 PURITY BASED ON TRACE METALS ANALYSIS
 APPROVED OCTOBER 10, 1997 JSB