Product Name
Ethyl (S)-(+)-mandelate 99%, optical purity ee: 99% (GLC)
Product Number
309974
Product Brand
aldrich
CAS Number
13704-09-1
Molecular Weight
180.20
Test
Specification
Appearance (Color)White
Appearance (Form)Conforms to Requirements
 Solid, Powder or Crystals
Infrared spectrumConforms to Structure
Optical Rotation133 - 137
 Degrees C=3%, CHCl3
Enantiomeric Excess≥98.5 %
Purity (GC)≥98.5 %
Solubility (Turbidity)Clear
 50mg/ml, CHCl3
Solubility (Color)Colorless to Very Faint Yellow