Product Name
Barium bis(2,2,6,6-tetramethyl-3,5-heptanedionate) hydrate
Product Number
362948
Product Brand
aldrich
CAS Number
17594-47-7
Molecular Weight
503.86 (anhydrous basis)
Test
Specification
APPEARANCEWHITE POWDER
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GRAVIMETRIC ASSAY26.3% BA (MINIMUM) WATER CORRECTED (WITH
 SULFURIC ACID)
TITRATION1.8% H2O (MINIMUM)
 APPROVED JULY 19, 2005 RJM