Product Name
2,2′-(o-Phenylenedioxy)diethanol 97%
Product Number
374687
Product Brand
aldrich
CAS Number
10234-40-9
Molecular Weight
198.22
Test
Specification
APPEARANCEWHITE TO BROWN POWDER, CRYSTALS, CRYSTALLINE
 POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 58.4%-62.7%
GAS LIQUID96.5% (MINIMUM)
SOLUBILITY50MG/ML METHANOL; CLEAR TO SLIGHTLY HAZY,
 COLORLESS TO YELLOW SOLUTION
 APPROVED DECEMBER 5, 2007 RJM