Product Name
5-Chloro-2-methylbenzoxazole 98%
Product Number
404500
Product Brand
aldrich
CAS Number
19219-99-9
Molecular Weight
167.59
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER, CRYSTALS,
 CRYSTALLINE POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION97.5%-102.5%(WITH AGNO3 AFTER O2 COMBUSTION)
HIGH PRESSURE LIQUID97.5% (MINIMUM)
 APPROVED FEBRUARY 13, 2006 RJM