Product Name
Poly(ethylene glycol-ran-propylene glycol) monobutyl ether average Mn ~1,700
Product Number
438170
Product Brand
aldrich
CAS Number
9038-95-3
Molecular Weight
Test
Specification
APPEARANCEVISCOUS COLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION0.13% H2O (MAXIMUM) *
COLOR TEST50 APHA (MAXIMUM) *
PH5.5 - 7.5 (C=10%, H2O @ 25 DEGREES C) *
 * SUPPLIER SPECIFICATION
 REVISED JULY 24, 2001 JSB