Product Name
2,2′-Thenil 98%
Product Number
454389
Product Brand
aldrich
CAS Number
7333-07-5
Molecular Weight
222.28
Test
Specification
APPEARANCE (COLOR)White to Yellow
APPEARANCE (FORM)Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
PURITY (HPLC AREA %)≥ 97.5 %
CARBON CONTENT52.7 - 55.4 %
SULFUR CONTENT28.1 - 29.6 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE