Product Name
(Aminomethyl)polystyrene 200-400 mesh, extent of labeling: 4.0 mmol/g loading, 2 % cross-linked
Product Number
473677
Product Brand
aldrich
CAS Number
Molecular Weight
Test
Specification
Appearance (Color)Tan
Appearance (Form)Powder
Carbon%
Nitrogen4.2 - 7.0 %
Loss on Drying%
 50 Degrees Celsius, 4 Hours
Miscellaneous AssayPowder
 Swelling Volume ml/g (Tetrahydrofuran)