Product Name
2-Aminoethyl methacrylate hydrochloride contains ~500 ppm phenothiazine as stabilizer, 90%
Product Number
516155
Product Brand
aldrich
CAS Number
2420-94-2
Molecular Weight
165.62
Test
Specification
STABILIZERcontains ~500ppm Phenothiazine
APPEARANCE (COLOR)White to Green to Beige
APPEARANCE (FORM)Powder
ARGENTOMETR. TITRATION89.0 - 111.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE