Product Name
(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)-1-propanol 98%
Product Number
548979
Product Brand
aldrich
CAS Number
56571-91-6
Molecular Weight
205.30
Test
Specification
APPEARANCEWHITE TO YELLOW SOLID, POWDER OR CRYSTALS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 74.1%-78.0%
 NITROGEN 6.6%- 7.0%
OPTICAL ROTATION-15(+/-2) DEGREES (C=2%, CHCL3)
 APPROVED NOVEMBER 17, 2008 RJM