Product Name
2-(Diphenylphosphino)terephthalic acid 1-methyl 4-pentafluorophenyl diester 97%
Product Number
679011
Product Brand
aldrich
CAS Number
Molecular Weight
530.38
Test
Specification
APPEARANCEWHITE TO TAN SOLID, POWDER, CRYSTALS,
 CRYSTALLINE POWDER, AND/OR WITH CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 59.0% - 63.3%
HIGH PRESSURE LIQUID96.5% (MINIMUM)
 APPROVED NOVEMBER 2, 2006 DJP