Product Name
Methyl phenyl sulfone ≥98.0% (GC)
Product Number
68742
Product Brand
aldrich
CAS Number
3112-85-4
Molecular Weight
156.20
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Powder or Crystals
PURITY (GC AREA %)≥ 98.0 %
MELTING POINT85 - 87 C
CARBON CONTENT53.83 % (THEORY)
HYDROGEN CONTENT5.16 % (THEORY)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE