Product Name
(S,S)-2,2′-Bipyrrolidine D-tartrate trihydrate 99%
Product Number
688746
Product Brand
aldrich
CAS Number
136937-03-6
Molecular Weight
344.36
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
PURITY (GC AREA %)≥ 98.5 %
ENANTIOMERIC EXCESS≥ 99.5 %
CARBON CONTENT41.85 % (Theory)
HYDROGEN CONTENT8.20 % (Theory)
NITROGEN CONTENT8.13 % (Theory)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE