Product Name
((4-Trifluoromethyl)phenyl)di-tert-butylphosphine technical grade, 85%
Product Number
695246
Product Brand
aldrich
CAS Number
1228182-34-0
Molecular Weight
290.30
Test
Specification
Appearance (Form)Conforms to Requirements
 Crystals or Crystalline Powder
Infrared spectrumConforms to Structure
Carbon51.2 - 73.0 %
Purity (GC)≥82.5 %