Product Name
MePhos 97%
Product Number
695262
Product Brand
aldrich
CAS Number
251320-86-2
Molecular Weight
364.50
Test
Specification
Appearance (Form)Conforms to Requirements
 Solid, Powder, Crystals, Crystalline Powder and/or
 Chunks
Infrared spectrumConforms to Structure
Purity (GC)≥96.5 %