Product Name
Pseudoionone technical, mixture of isomers, ≥90% (GC)
Product Number
82536
Product Brand
aldrich
CAS Number
141-10-6
Molecular Weight
192.30
Test
Specification
APPEARANCE (COLOR)Yellow to Very Dark Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 90 %
REMARKS ON GCMixture of Isomers
DENSITY D20/40.890 - 0.910
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE