Product Name
1,4-Dimethylnaphthalene 95%
Product Number
d170305
Product Brand
aldrich
CAS Number
571-58-4
Molecular Weight
156.22
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow and Colorless to Brown and Colorless to Red
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 94.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE