Product Name
trans-1,4-Diphenyl-2-butene-1,4-dione 96%
Product Number
d33209
Product Brand
aldrich
CAS Number
959-28-4
Molecular Weight
236.27
Test
Specification
APPEARANCEYELLOW POWDER, CRYSTALS OR CRYSTALLINE
 POWDER
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 77.7%-85.0%
GAS LIQUID95.5% (MINIMUM)
 APPROVED DECEMBER 6, 2004