Product Name
1,3-Dichlorobutane 99%
Product Number
d59003
Product Brand
aldrich
CAS Number
1190-22-3
Molecular Weight
127.01
Test
Specification
Appearance (Color)Colorless
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (GC)≥98.5 %