Product Name
3-Methylindole 98%
Product Number
m51458
Product Brand
aldrich
CAS Number
83-34-1
Molecular Weight
131.17
Test
Specification
APPEARANCE (COLOR)White to Light Yellow
APPEARANCE (FORM)Powder or Crystals or Flakes
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE