Product Name
Thioacetic acid 96%
Product Number
t30805
Product Brand
aldrich
CAS Number
507-09-5
Molecular Weight
76.12
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 95.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE