Product Name
Mineral oil standard mixture Type A and B for EN ISO 9377-2 analytical standard, ~10 mg/mL in heptane (exact content on the label)
Product Number
18602
Product Brand
sial
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)CORRESPONDS TO STANDARD
DENSITY D20/40.685 - 0.687
REFRACTIVE INDEX N20/D1.388 - 1.390
EXPIRY PERIOD24 MONTHS