Product Name
N-(Diphenylmethylene)glycine ethyl ester 98%
Product Number
222542
Product Brand
sial
CAS Number
69555-14-2
Molecular Weight
267.32
Test
Specification
APPEARANCE (COLOR)White to Yellow
APPEARANCE (FORM)Powder or Crystals or Chunk(s)
SOLUBILITY (COLOR)Colorless to Faint Yellow
SOLUBILITY (TURBIDITY)Clear to Very Slightly Hazy
SOLUBILITY (METHOD)50MG/ML ETHANOL
CARBON CONTENT74.5 - 78.3 %
NITROGEN CONTENT4.9 - 5.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE