Product Name
2-Acetylpyrrole analytical standard
Product Number
30642
Product Brand
sial
CAS Number
1072-83-9
Molecular Weight
109.13
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (GC AREA %)≥ 98.5 %
WATER≤ 0.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS