Product Name
Ochratoxin A solution 10 μg/mL in acetonitrile, analytical standard
Product Number
34037
Product Brand
sial
CAS Number
303-47-9
Molecular Weight
403.81
Test
Specification
CONCENTRATION COMP1~ 10 ug/ml
MEASURING TOLERANCE P±≤ 1.00 ug/ml
SOLVENTACETONITRILE
PURITY (HPLC) COMP1≥ 98.0 %
MEASURING TOLERANCE P± COMP1≤ 2.0 %