Product Name
Cinmethylin PESTANAL®, analytical standard
Product Number
34237
Product Brand
sial
CAS Number
87818-31-3
Molecular Weight
274.40
Test
Specification
APPEARANCE (COLOR)Very Faint Yellow to Dark Yellow
APPEARANCE (FORM)Liquid or Viscous Liquid
PURITY (HPLC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.5098 - 1.5138
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD48 MONTHS