Product Name
Cloquintocet PESTANAL®, analytical standard
Product Number
34239
Product Brand
sial
CAS Number
88349-88-6
Molecular Weight
237.64
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals or Fibers or Solid
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT220 - 225 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS