Product Name
Sulfur PESTANAL®, analytical standard
Product Number
36576
Product Brand
sial
CAS Number
7704-34-9
Molecular Weight
32.07
Test
Specification
APPEARANCE (COLOR)Faint Yellow to Very Dark Yellow and Faint Beige to Dark Beige and Faint Brown to Brown
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) NAOH 0.1M95 - 106 %
MELTING POINT116 - 122 C
SULFUR CONTENT (S/C-ANALYSIS)95 - 106 %
EXPIRY PERIOD60 MONTHS