Product Name
3,4-Dihydroxybenzaldehyde purum, ≥97.0% (HPLC)
Product Number
37520
Product Brand
sial
CAS Number
139-85-5
Molecular Weight
138.12
Test
Specification
APPEARANCE (COLOR)White to Yellow and Faint Beige to Beige and Faint Brown to Brown and Faint Grey to Grey
APPEARANCE (FORM)Powder
FREE CARBOXYLIC ACID (T)≤ 3.0 %
PURITY (HPLC AREA %)≥ 97.0 %
MELTING POINT150 - 155 C
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE