Product Name
Chlorflurenol PESTANAL®, analytical standard
Product Number
45721
Product Brand
sial
CAS Number
2464-37-1
Molecular Weight
260.67
Test
Specification
PURITY (HPLC AREA %)≥ 90.0 %
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS