Product Name
[(3R)-3-Hydroxy-cis-tetradec-9-enoyl]-L-carnitine analytical standard
Product Number
52018
Product Brand
sial
CAS Number
Molecular Weight
385.54
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 84.0 %
IDENTIFICATION (LC-MS)CORRESPONDS TO REQUIREMENTS
SPECIFIC ROTATION (20/D)-20.0 ± 2.0 Degrees
CONCENTRATIONc=0.1 in Methanol
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS