Product Name
Hexadecyltrimethylammonium bromide ≥96.0% (AT)
Product Number
52370
Product Brand
sial
CAS Number
57-09-0
Molecular Weight
364.45
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Powder with Chunk(s)
ARGENTOMETR. TITRATION96.0 - 104.0 %
CARBON CONTENT62.62 % (THEORY)
HYDROGEN CONTENT11.62 % (THEORY)
NITROGEN CONTENT3.84 % (THEORY)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD42 MONTHS