Product Name
Methyl dihydrojasmonate, mixture of cis and trans analytical standard
Product Number
59677
Product Brand
sial
CAS Number
24851-98-7
Molecular Weight
226.31
Test
Specification
APPEARANCE (COLOR)Colorless to Almost Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 96.0 %
REMARKS ON GCMixture of Cis and Trans Isomers
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS