Product Name
Benalaxyl reference material
Product Number
62266
Product Brand
sial
CAS Number
71626-11-4
Molecular Weight
325.40
Test
Specification
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
EXPIRY PERIOD24 MONTHS