Product Name
1-Naphthol reference material
Product Number
69442
Product Brand
sial
CAS Number
90-15-3
Molecular Weight
144.17
Test
Specification
Assay (QNMR)*Report certified value and uncertainty (acc. to ISO17025)
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (GC AREA %)≥ 97.0 %
EXPIRY PERIOD24 MONTHS