Product Name
O-Acetyl-L-carnitine hydrochloride analytical standard
Product Number
92107
Product Brand
sial
CAS Number
5080-50-2
Molecular Weight
239.70
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
ARGENTOMETR. TITRATION96.0 - 104.0 %
PURITY (HPLC AREA %)≥ 96.0 %
SPECIFIC ROTATION (20/D)-29.0 - -25.0 Degrees
CONCENTRATIONc=1 in water
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS