Product Name
N-Heptanoylglycine analytical standard
Product Number
93539
Product Brand
sial
CAS Number
23783-23-5
Molecular Weight
187.24
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals or Fibers
PURITY (HPLC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS