Product Name
N-Cinnamoylglycine analytical standard
Product Number
98956
Product Brand
sial
CAS Number
16534-24-0
Molecular Weight
205.21
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS