Product Name
2-Propanol suitable for HPLC, 99.9%
Product Number
34863
Product Brand
sigald
CAS Number
67-63-0
Molecular Weight
60.10
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
COLOR (IN APHA)≤ 10 APHA
PURITY (GC AREA %)≥ 99.85 %
WATER (COULOMETR.)≤ 0.05 %
RESIDUE (EVAPORATION)≤ 0.0003 %
UV-ABSORBANCE400nm ≤ 0.01
UV-ABSORBANCE260nm ≤ 0.01
UV-ABSORBANCE230nm ≤ 0.10
UV-ABSORBANCE220nm ≤ 0.20
UV-ABSORBANCE210nm ≤ 0.70
UV-ABSORBANCE205nm ≤ 1.0
ACIDITY≤ 0.001 % as Acetic Acid
RECOMMENDED RETEST PERIOD42 Months