Product Name
Zinc chloride solution 0.5 M in THF
Product Number
366374
Product Brand
sigald
CAS Number
7646-85-7
Molecular Weight
136.30
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
TITRATION (METHOD)WITH AgNO3
TITRATION (MOLARITY)0.47 - 0.60 mol/l