Product Name
5(6)-Carboxy-2′,7′-dichlorofluorescein BioReagent, suitable for fluorescence, ≥95% (TLC)
Product Number
21882
Product Brand
sigma
CAS Number
111843-78-8
Molecular Weight
445.21
Test
Specification
APPEARANCE (COLOR)Light Orange to Dark Orange
APPEARANCE (FORM)Powder or Crystals
PURITY (TLC AREA %)≥ 95.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
LUMINESCENCECORRESPONDS TO REQUIREMENTS
EMISSION MAXIMUM529 ± 5 nm